4-Bromo-2-((4-methoxybenzyl)oxy)-1-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

60696-1G 60696-5G 60696-25G
77341-044EA 66.95 USD
77341-044 77367-196 77379-290
4-Bromo-2-((4-methoxybenzyl)oxy)-1-methylbenzene ≥95%
4-Bromo-2-((4-methoxybenzyl)oxy)-1-methylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO₂
MW: 307.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30296478
CAS Number: 1207956-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR