2-Bromo-1-((4-methoxybenzyl)oxy)-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

60694-500MG 60694-1G 60694-5G
77336-432EA 387.28 USD
77336-432 77355-212 77379-286
2-Bromo-1-((4-methoxybenzyl)oxy)-4-methylbenzene ≥97%
2-Bromo-1-((4-methoxybenzyl)oxy)-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO₂
MW: 307.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11116843
CAS Number: 1036593-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR