2-Fluoro-1,3-diiodo-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

60691-500MG 60691-1G 60691-25G 60691-5G
77330-996EA 292.52 USD
77330-996 77355-210 77388-736 77379-284
2-Fluoro-1,3-diiodo-5-methylbenzene ≥97%
2-Fluoro-1,3-diiodo-5-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅FI₂
MW: 361.922 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28791809
CAS Number: 1806355-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR