2-Chloro-4-fluoro-3-(methoxymethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

60689-500MG 60689-1G 60689-5G
77339-194EA 562.38 USD
77339-194 77355-208 77379-282
2-Chloro-4-fluoro-3-(methoxymethyl)benzaldehyde ≥97%
2-Chloro-4-fluoro-3-(methoxymethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759888
CAS Number: 2764732-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR