1,2,5-Tribromo-3-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

60684-500MG 60684-1G 60684-5G 60684-25G
77330-992EA 278.1 USD
77330-992 77355-202 77367-192 77388-734
1,2,5-Tribromo-3-nitrobenzene ≥97%
1,2,5-Tribromo-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD18400067
CAS Number: 116529-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR