(5-Chloro-3-ethoxy-2-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

60678-500MG 60678-1G 60678-5G 60678-25G
77330-988EA 147.29 USD
77330-988 77345-230 77367-188 77388-730
(5-Chloro-3-ethoxy-2-fluorophenyl)(methyl)sulfane ≥97%
(5-Chloro-3-ethoxy-2-fluorophenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₉H₁₀ClFOS
MW: 220.692 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759882
CAS Number: 2705712-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR