Methyl 5-chloro-3-ethoxy-2-fluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

60674-500MG 60674-1G 60674-5G 60674-25G
77330-980EA 230.72 USD
77330-980 77355-194 77367-180 77388-724
Methyl 5-chloro-3-ethoxy-2-fluorobenzoate ≥95%
Methyl 5-chloro-3-ethoxy-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₃
MW: 232.636 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759878
CAS Number: 2387201-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR