5-Chloro-3-ethoxy-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

60673-500MG 60673-1G 60673-5G 60673-25G
77330-978EA 152.44 USD
77330-978 77345-466 77367-178 77388-722
5-Chloro-3-ethoxy-2-fluorobenzoic acid ≥97%
5-Chloro-3-ethoxy-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈ClFO₃
MW: 218.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759877
CAS Number: 2384573-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR