1-(Benzyloxy)-3-bromo-5-chloro-2-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

60670-500MG 60670-1G 60670-5G 60670-25G
77330-974EA 243.08 USD
77330-974 77355-192 77367-174 77388-720
1-(Benzyloxy)-3-bromo-5-chloro-2-fluorobenzene ≥97%
1-(Benzyloxy)-3-bromo-5-chloro-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉BrClFO
MW: 315.565 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759874
CAS Number: 2901098-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR