1-Bromo-5-chloro-2-fluoro-3-isopropoxybenzene ≥95%

Supplier: AOBChem USA

Warning

60669-500MG 60669-1G 60669-5G 60669-25G
77330-972EA 192.61 USD
77330-972 77355-190 77367-172 77388-718
1-Bromo-5-chloro-2-fluoro-3-isopropoxybenzene ≥95%
1-Bromo-5-chloro-2-fluoro-3-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrClFO
MW: 267.523 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29043861
CAS Number: 1881332-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR