4-Phenyl-1-naphthaldehyde ≥97%

Supplier: AOBChem USA

Warning

60651-500MG 60651-1G 60651-5G 60651-25G
77330-966EA 184.37 USD
77330-966 77347-168 77367-166 77388-712
4-Phenyl-1-naphthaldehyde ≥97%
4-Phenyl-1-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₇H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD17012503
CAS Number: 133550-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR