1,2,4-Tribromonaphthalene ≥95%

Supplier: AOBChem USA

Warning

60627-500MG 60627-1G 60627-5G 60627-25G
77330-962EA 120.51 USD
77330-962 77343-772 77367-160 77379-270
1,2,4-Tribromonaphthalene ≥95%
1,2,4-Tribromonaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₅Br₃
Storage Temperature: Ambient
MDL Number: MFCD34759855
CAS Number: 64725-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR