3-Acetyl-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

60626-500MG 60626-1G 60626-5G
77340-340EA 908.46 USD
77340-340 77355-184 77379-268
3-Acetyl-5-(trifluoromethyl)benzaldehyde ≥97%
3-Acetyl-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD22544358
CAS Number: 1393534-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR