tert-Butyl 6-bromo-2,3-dichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

60619-500MG 60619-1G 60619-5G
77335-438EA 353.29 USD
77335-438 77355-180 77379-266
tert-Butyl 6-bromo-2,3-dichlorobenzoate ≥97%
tert-Butyl 6-bromo-2,3-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrCl₂O₂
MW: 326.014 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759854
CAS Number: 2757730-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR