Ethyl 6-bromo-2,3-dichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

60618-500MG 60618-1G 60618-5G
77330-958EA 328.57 USD
77330-958 77355-178 77379-264
Ethyl 6-bromo-2,3-dichlorobenzoate ≥97%
Ethyl 6-bromo-2,3-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrCl₂O₂
MW: 297.961 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759853
CAS Number: 2167328-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR