1,5-Dibromo-3-chloro-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

60617-500MG 60617-1G 60617-5G 60617-25G
77330-956EA 128.75 USD
77330-956 77344-268 77367-156 77379-262
1,5-Dibromo-3-chloro-2-methylbenzene ≥97%
1,5-Dibromo-3-chloro-2-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Br₂Cl
MW: 284.376 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759852
CAS Number: 2385656-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR