1,2,5-Tribromo-4-iodo-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

60612-500MG 60612-1G 60612-5G 60612-25G
77330-952EA 152.44 USD
77330-952 77345-464 77367-152 77388-706
1,2,5-Tribromo-4-iodo-3-methylbenzene ≥97%
1,2,5-Tribromo-4-iodo-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₄Br₃I
MW: 454.723 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759851
CAS Number: 2764728-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR