1-(2-Chloro-4,5-dimethoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

60606-500MG 60606-1G 60606-5G 60606-25G
77330-950EA 149.35 USD
77330-950 77345-314 77367-150 77388-704
1-(2-Chloro-4,5-dimethoxyphenyl)ethanone ≥97%
1-(2-Chloro-4,5-dimethoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₃
Storage Temperature: Ambient
MDL Number: MFCD07339247
CAS Number: 161199-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR