4-(Benzyloxy)-2-chloro-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

60600-500MG 60600-1G 60600-5G
77330-946EA 321.36 USD
77330-946 77355-172 77379-256
4-(Benzyloxy)-2-chloro-5-methoxybenzaldehyde ≥97%
4-(Benzyloxy)-2-chloro-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO₃
MW: 276.715 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02379931
CAS Number: 225110-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR