2-Chloro-4-ethoxy-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

60579-500MG 60579-1G 60579-5G 60579-25G
77330-944EA 265.74 USD
77330-944 77355-166 77367-146 77388-700
2-Chloro-4-ethoxy-5-methoxybenzoic acid ≥97%
2-Chloro-4-ethoxy-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₄
MW: 230.645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28247066
CAS Number: 1794756-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR