1-Bromo-2-chloro-4-(difluoromethoxy)-5-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

60575-500MG 60575-1G 60575-5G
77336-108EA 375.95 USD
77336-108 77355-162 77379-250
1-Bromo-2-chloro-4-(difluoromethoxy)-5-methoxybenzene ≥97%
1-Bromo-2-chloro-4-(difluoromethoxy)-5-methoxybenzene

Precaución: for Research Use Only

Formula: C₈H₆BrClF₂O₂
MW: 287.486 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759833
CAS Number: 2091517-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR