1-(Benzyloxy)-4-bromo-5-chloro-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

60573-500MG 60573-1G 60573-5G
77337-386EA 413.03 USD
77337-386 77355-160 77379-248
1-(Benzyloxy)-4-bromo-5-chloro-2-methoxybenzene ≥97%
1-(Benzyloxy)-4-bromo-5-chloro-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrClO₂
MW: 327.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759831
CAS Number: 2764733-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR