1-Bromo-2-chloro-4-ethoxy-5-methoxybenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
60572-500MG
60572-1G
60572-5G
77340-320EA
900.22
USD
77340-320
77355-158
77379-246
1-Bromo-2-chloro-4-ethoxy-5-methoxybenzene ≥97%
1-Bromo-2-chloro-4-ethoxy-5-methoxybenzene
Precaución: for Research Use Only
Formula:
C₉H₁₀BrClO₂ MW: 265.531 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34759830 CAS Number: 2386542-40-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...