2-(5-Bromo-2-fluoro-3-methoxyphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

60569-500MG 60569-1G 60569-5G
77335-604EA 360.5 USD
77335-604 77355-154 77379-244
2-(5-Bromo-2-fluoro-3-methoxyphenyl)propan-2-ol ≥97%
2-(5-Bromo-2-fluoro-3-methoxyphenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO₂
MW: 263.103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759828
CAS Number: 1783404-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR