1-(5-Bromo-2-fluoro-3-methoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

60567-500MG 60567-1G 60567-5G
77335-702EA 363.59 USD
77335-702 77355-152 77379-242
1-(5-Bromo-2-fluoro-3-methoxyphenyl)ethanone ≥97%
1-(5-Bromo-2-fluoro-3-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MW: 247.061 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759826
CAS Number: 1784314-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR