5-Bromo-2-fluoro-1-methoxy-3-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

60566-500MG 60566-1G 60566-5G 60566-25G
77330-940EA 187.46 USD
77330-940 77355-150 77367-142 77388-696
5-Bromo-2-fluoro-1-methoxy-3-(methylsulfonyl)benzene ≥97%
5-Bromo-2-fluoro-1-methoxy-3-(methylsulphonyl)benzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO₃S
MW: 283.115 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759825
CAS Number: 2755719-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR