Ethyl 4-ethyl-2-formylbenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
60558-500MG
60558-1G
60558-5G
60558-25G
77330-936EA
226.6
USD
77330-936
77355-146
77367-138
77388-692
Ethyl 4-ethyl-2-formylbenzoate ≥97%
Ethyl 4-ethyl-2-formylbenzoate
Precaución: for Research Use Only
Formula:
C₁₂H₁₄O₃ Storage Temperature: Ambient |
MDL Number:
MFCD34759819 CAS Number: 2755716-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...