1-Butyl-4-chloro-3,5-dimethyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

60546-1G 60546-5G 60546-25G
77341-348EA 82.4 USD
77341-348 77367-136 77379-238
1-Butyl-4-chloro-3,5-dimethyl-1H-pyrazole ≥97%
1-Butyl-4-chloro-3,5-dimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₁₅ClN₂
MW: 186.682 g/mol
Storage Temperature: Ambient
CAS Number: 1205839-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR