4-Chloro-3,5-dimethyl-1-propyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

60545-500MG 60545-1G 60545-25G 60545-5G
77330-934EA 303.85 USD
77330-934 77355-142 77388-690 77379-236
4-Chloro-3,5-dimethyl-1-propyl-1H-pyrazole ≥97%
4-Chloro-3,5-dimethyl-1-propyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₈H₁₃ClN₂
MW: 172.655 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24153914
CAS Number: 1205195-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR