4-Chloro-1-ethyl-3,5-dimethyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

60539-500MG 60539-1G 60539-5G 60539-25G
77330-928EA 174.07 USD
77330-928 77346-458 77367-130 77388-688
4-Chloro-1-ethyl-3,5-dimethyl-1H-pyrazole ≥97%
4-Chloro-1-ethyl-3,5-dimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₇H₁₁ClN₂
Storage Temperature: Ambient
MDL Number: MFCD12026909
CAS Number: 1171799-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR