3-Fluoro-2-iodo-4-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

60536-500MG 60536-1G 60536-5G 60536-25G
77330-924EA 256.47 USD
77330-924 77355-140 77367-126 77388-684
3-Fluoro-2-iodo-4-methoxyphenol ≥97%
3-Fluoro-2-iodo-4-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆FIO₂
MW: 268.024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759813
CAS Number: 2385401-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR