3-Fluoro-4-methoxy-2-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

60534-500MG 60534-1G 60534-5G
77337-106EA 407.88 USD
77337-106 77355-138 77379-230
3-Fluoro-4-methoxy-2-(methylthio)phenol ≥97%
3-Fluoro-4-methoxy-2-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₈H₉FO₂S
MW: 188.219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759811
CAS Number: 2705650-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR