Methyl 2-fluoro-6-hydroxy-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

60530-500MG 60530-1G 60530-25G 60530-5G
77330-922EA 301.79 USD
77330-922 77355-136 77388-682 77379-228
Methyl 2-fluoro-6-hydroxy-3-methoxybenzoate ≥97%
Methyl 2-fluoro-6-hydroxy-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₄
Storage Temperature: Ambient
MDL Number: MFCD34759808
CAS Number: 2091971-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR