6-(Benzyloxy)-2-fluoro-3-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

60520-500MG 60520-1G 60520-5G 60520-25G
77330-914EA 128.75 USD
77330-914 77344-266 77367-118 77379-224
6-(Benzyloxy)-2-fluoro-3-methoxybenzoic acid ≥97%
6-(Benzyloxy)-2-fluoro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₄
Storage Temperature: Ambient
MDL Number: MFCD34759799
CAS Number: 2807465-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR