1-Ethoxy-3-fluoro-2-iodo-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

60509-500MG 60509-1G 60509-5G
77338-388EA 484.1 USD
77338-388 77355-132 77379-216
1-Ethoxy-3-fluoro-2-iodo-4-methoxybenzene ≥97%
1-Ethoxy-3-fluoro-2-iodo-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀FIO₂
MW: 296.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759788
CAS Number: 2384445-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR