2-Fluoro-4-isopropoxy-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

60500-1G 60500-5G 60500-25G
77340-766EA 60.77 USD
77340-766 77367-106 77379-206
2-Fluoro-4-isopropoxy-1-methoxybenzene ≥97%
2-Fluoro-4-isopropoxy-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO₂
MW: 184.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23113584
CAS Number: 1369847-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR