4-Ethoxy-2-fluoro-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

60499-500MG 60499-1G 60499-5G
77330-904EA 318.27 USD
77330-904 77355-124 77379-204
4-Ethoxy-2-fluoro-1-methoxybenzene ≥97%
4-Ethoxy-2-fluoro-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759779
CAS Number: 1881077-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR