2-Chloro-3-fluoro-1,4-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

60498-500MG 60498-1G 60498-5G 60498-25G
77330-902EA 230.72 USD
77330-902 77355-122 77367-104 77388-674
2-Chloro-3-fluoro-1,4-dimethoxybenzene ≥97%
2-Chloro-3-fluoro-1,4-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈ClFO₂
MW: 190.599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751445
CAS Number: 1807122-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR