(2-Fluoro-3,6-dimethoxyphenyl)(methyl)sulphane ≥97%

Supplier: AOBChem USA

Warning

60497-500MG 60497-1G 60497-5G 60497-25G
77330-900EA 120.51 USD
77330-900 77343-768 77367-102 77379-202
(2-Fluoro-3,6-dimethoxyphenyl)(methyl)sulphane ≥97%
(2-Fluoro-3,6-dimethoxyphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂S
Storage Temperature: Ambient
MDL Number: MFCD28793159
CAS Number: 1805064-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR