1-Bromo-4-fluoro-2,5-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

60495-500MG 60495-1G 60495-5G
77335-866EA 367.71 USD
77335-866 77355-118 77379-198
1-Bromo-4-fluoro-2,5-dimethoxybenzene ≥97%
1-Bromo-4-fluoro-2,5-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28738432
CAS Number: 1783644-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR