(6-Bromo-2-fluoro-3-propoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

60481-500MG 60481-1G 60481-5G 60481-25G
77330-894EA 202.91 USD
77330-894 77355-110 77367-098 77388-670
(6-Bromo-2-fluoro-3-propoxyphenyl)(methyl)sulfane ≥97%
(6-Bromo-2-fluoro-3-propoxyphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFOS
MW: 279.169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759771
CAS Number: 2757731-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR