1-Bromo-2-ethoxy-3-fluoro-4-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

60478-500MG 60478-1G 60478-5G 60478-25G
77330-890EA 126.69 USD
77330-890 77344-216 77367-094 77379-190
1-Bromo-2-ethoxy-3-fluoro-4-iodobenzene ≥97%
1-Bromo-2-ethoxy-3-fluoro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₇BrFIO
Storage Temperature: Ambient
MDL Number: MFCD34759768
CAS Number: 2383867-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR