4-Chloro-2-ethyl-3-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

60460-500MG 60460-1G 60460-5G
77340-362EA 931.12 USD
77340-362 77355-106 77379-186
4-Chloro-2-ethyl-3-fluorobenzoic acid ≥97%
4-Chloro-2-ethyl-3-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759757
CAS Number: 2711576-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR