1-(4,5-Difluoro-2-methoxyphenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
60455-500MG
60455-1G
60455-5G
77330-888EA
316.21
USD
77330-888
77355-104
77379-184
1-(4,5-Difluoro-2-methoxyphenyl)ethanone ≥97%
1-(4,5-Difluoro-2-methoxyphenyl)ethanone
Precaución: for Research Use Only
Formula:
C₉H₈F₂O₂ MW: 186.155 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11553405 CAS Number: 1256477-94-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...