1-(3,5-Dimethoxyphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

60437-500MG 60437-1G 60437-5G 60437-25G
77330-886EA 175.1 USD
77330-886 77346-492 77367-086 77388-668
1-(3,5-Dimethoxyphenyl)ethanol ≥95%
1-(3,5-Dimethoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD19601840
CAS Number: 14950-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR