2,5-Dichloro-1,3-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

60433-500MG 60433-1G 60433-5G 60433-25G
77330-884EA 264.71 USD
77330-884 77355-102 77367-084 77388-666
2,5-Dichloro-1,3-difluorobenzene ≥97%
2,5-Dichloro-1,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₂F₂
MW: 182.983 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922631
CAS Number: 36611-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR