2,2-Dichloro-1-(4-chloro-3,5-dimethylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

60427-500MG 60427-1G 60427-5G 60427-25G
77330-882EA 149.35 USD
77330-882 77345-312 77367-082 77388-664
2,2-Dichloro-1-(4-chloro-3,5-dimethylphenyl)ethanol ≥95%
2,2-Dichloro-1-(4-chloro-3,5-dimethylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₁Cl₃O
MW: 253.553 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759751
CAS Number: 2755724-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR