(4-Bromo-1-phenyl-1H-pyrazol-5-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

60393-500MG 60393-1G 60393-5G
77338-158EA 466.59 USD
77338-158 77355-100 77379-174
(4-Bromo-1-phenyl-1H-pyrazol-5-yl)methanol ≥97%
(4-Bromo-1-phenyl-1H-pyrazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂O
MW: 253.095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759724
CAS Number: 1499994-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR