4-Bromo-1-phenyl-1H-pyrazole-5-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

60392-500MG 60392-1G 60392-25G 60392-5G
77330-880EA 296.64 USD
77330-880 77355-098 77388-662 77379-172
4-Bromo-1-phenyl-1H-pyrazole-5-carbaldehyde ≥97%
4-Bromo-1-phenyl-1H-pyrazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇BrN₂O
MW: 251.079 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28401399
CAS Number: 1507703-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR