Ethyl 2,6-difluoro-4-iodobenzoate ≥97%

Supplier: AOBChem USA

Warning

60388-500MG 60388-1G 60388-25G 60388-5G
77330-876EA 292.52 USD
77330-876 77355-094 77388-658 77379-170
Ethyl 2,6-difluoro-4-iodobenzoate ≥97%
Ethyl 2,6-difluoro-4-iodobenzoate

Precaución: for Research Use Only

Formula: C₉H₇F₂IO₂
MW: 312.052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31707076
CAS Number: 2385126-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR